3-Hydroxy-1-adamantane carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxyadamantane-1-carboxylic acid, 3-Hydroxy-1-adamantanecarboxylic acid

A353240-100G A353240-1G A353240-25G A353240-5G A353240-500G A353240-1000G
77278-188EA 76 USD
77278-188 77278-190 77278-192 77278-194 77748-696 77748-695
3-Hydroxy-1-adamantane carboxylic acid 97%
3-Hydroxy-1-adamantane carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃
MDL Number: MFCD00193929
CAS Number: 42711-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR