Chloro(1,5-cyclooctadiene)copper(I) dimer 98%

Supplier: Ambeed
Warning

A353074-250MG A353074-1G A353074-100MG
77263-888EA 53 USD
77263-888 77572-038 77748-694
Chloro(1,5-cyclooctadiene)copper(I) dimer 98%
Chloro(1,5-cyclooctadiene)copper(I) dimer

Precaución: for Research Use Only

Formula: C₁₆H₁₆Cl₂Cu₂
MDL Number: MFCD23704409
CAS Number: 32717-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR