2-Amino-4,6-dimethoxybenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-amino-4,6-dimethoxybenzoic acid

A353041-10G A353041-1G A353041-250MG A353041-25G A353041-5G
77251-574EA 340.93 USD
77251-574 77251-576 77251-578 77251-580 77251-582
2-Amino-4,6-dimethoxybenzoic acid 98%
2-Amino-4,6-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄
MW: 197.1900 g/mol
MDL Number: MFCD09031989
CAS Number: 21577-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR