Methanesulfonato(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: Ambeed
Warning

A352841-100MG A352841-1G A352841-250MG A352841-5G
77251-566EA 10 USD
77251-566 77251-568 77251-570 77251-572
Methanesulfonato(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₄₀H₅₀NO₅PPdS
MDL Number: MFCD29905053
CAS Number: 1599466-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR