Chloro[1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]gold(I) 98%

Supplier: Ambeed
Warning

A352707-100MG A352707-250MG
77251-562EA 57 USD
77251-562 77251-564
Chloro[1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]gold(I) 98%
Chloro[1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene]gold(I)

Precaución: for Research Use Only

Formula: C₂₁H₂₆AuClN₂
MDL Number: MFCD29037185
CAS Number: 852445-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR