N3,N5,3'-Trimethyl-4'-(((2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-[1,1'-biphenyl]-3,5-dicarboxamide 98+%

Supplier: Ambeed
Warning

Synonyms: M4284

A352670-10MG A352670-25MG A352670-50MG A352670-5MG A352670-1MG
77263-880EA 42 USD
77263-880 77263-882 77263-884 77263-886 77616-546
N3,N5,3'-Trimethyl-4'-(((2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-[1,1'-biphenyl]-3,5-dicarboxamide 98+%
N3,N5,3'-Trimethyl-4'-(((2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-[1,1'-biphenyl]-3,5-dicarboxamide

Precaución: for Research Use Only

Formula: C₂₃H₂₈N₂O₈
MDL Number: MFCD32689437
CAS Number: 1373346-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR