2-Ethoxy-6-fluorobenzonitrile 96%

Supplier: Ambeed
Warning

A352590-10G A352590-1G
77263-876EA 35 USD
77263-876 77263-878
2-Ethoxy-6-fluorobenzonitrile 96%
2-Ethoxy-6-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₉H₈FNO
MDL Number: MFCD05664780
CAS Number: 119584-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR