4-Acetyl-4-(ethoxycarbonyl)heptane-1,7-dioic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Acetyl-4-(ethoxycarbonyl)heptanedioic acid, 4-Acetyl-4-(ethoxycarbonyl)pimelic acid, Ethyl 2,2-bis(2-carboxyethyl)acetoacetate, 3-Acetyl-3-(ethoxycarbonyl)pentane-1,5-dicarboxylic acid

A352587-100MG A352587-1G A352587-250MG
77290-674EA 25.75 USD
77290-674 77290-676 77290-678
4-Acetyl-4-(ethoxycarbonyl)heptane-1,7-dioic acid 98%
4-Acetyl-4-(ethoxycarbonyl)heptane-1,7-dioic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₇
MW: 274.267 g/mol
MDL Number: MFCD00052373
CAS Number: 72653-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR