(R,E)-5-([1,1'-Biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpent-2-enoic acid 97%

Supplier: Ambeed
Warning

A352438-10G A352438-1G A352438-5G
77240-016EA 37.08 USD
77240-016 77240-018 77240-020
(R,E)-5-([1,1'-Biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpent-2-enoic acid 97%
(R,E)-5-([1,1'-Biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpent-2-enoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₇NO₄
MW: 381.47 g/mol
MDL Number: MFCD28386950
CAS Number: 1012341-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR