α,α,α,2,4,6-Hexafluorotoluene 98%

Supplier: Ambeed
Danger

A352427-25G A352427-5G
77251-558EA 237 USD
77251-558 77251-560
α,α,α,2,4,6-Hexafluorotoluene 98%
α,α,α,2,4,6-Hexafluorotoluene

Precaución: for Research Use Only

Formula: C₇H₂F₆
MDL Number: MFCD00190109
CAS Number: 122030-04-0
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR