2,6-Dichloro-3-hydroxybenzonitrile 96%

Supplier: Ambeed
Danger

A352102-100MG A352102-250MG
77240-012EA 119 USD
77240-012 77240-014
2,6-Dichloro-3-hydroxybenzonitrile 96%
2,6-Dichloro-3-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO
MDL Number: MFCD01662155
CAS Number: 3336-34-3
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR