Diisobutyl terephthalate 97%

Supplier: Ambeed
Warning

A352091-100MG A352091-1G A352091-250MG
77240-006EA 6 USD
77240-006 77240-008 77240-010
Diisobutyl terephthalate 97%
Diisobutyl terephthalate

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₄
MDL Number: MFCD01682719
CAS Number: 18699-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR