(S)-3-Methyl-2-(2-oxotetrahydropyrimidin-1(2H)-yl)butanoic acid 97%

Supplier: Ambeed
Warning

A351735-25G A351735-5G
77251-554EA 46 USD
77251-554 77251-556
(S)-3-Methyl-2-(2-oxotetrahydropyrimidin-1(2H)-yl)butanoic acid 97%
(S)-3-Methyl-2-(2-oxotetrahydropyrimidin-1(2H)-yl)butanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O₃
MW: 200.23 g/mol
MDL Number: MFCD06797211
CAS Number: 192725-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR