Ethyl 3-(3-cyanophenyl)-3-oxopropanoate 97%

Supplier: Ambeed
Warning

A351673-1G A351673-250MG A351673-5G
77278-168EA 94.76 USD
77278-168 77278-170 77278-172
Ethyl 3-(3-cyanophenyl)-3-oxopropanoate 97%
Ethyl 3-(3-cyanophenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₃
MDL Number: MFCD06656737
CAS Number: 62088-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR