Naphthalen-2-ylmethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A351530-1G A351530-5G
77278-164EA 27.81 USD
77278-164 77278-166
Naphthalen-2-ylmethanamine hydrochloride 95%
Naphthalen-2-ylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClN
MW: 193.6700 g/mol
MDL Number: MFCD00137202
CAS Number: 2241-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR