2-Bromo-4,6-dimethylpyrimidine 98%

Supplier: Ambeed
Warning

A351365-100MG A351365-250MG A351365-5G
77263-862EA 63.86 USD
77263-862 77263-864 77263-866
2-Bromo-4,6-dimethylpyrimidine 98%
2-Bromo-4,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD09910233
CAS Number: 16879-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR