4-Fluoro-2-nitrophenol 96%

Supplier: Ambeed
Warning

A350743-100G A350743-10G A350743-25G A350743-5G
77290-660EA 63.86 USD
77290-660 77290-662 77290-664 77290-666
4-Fluoro-2-nitrophenol 96%
4-Fluoro-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄FNO₃
MDL Number: MFCD00042528
CAS Number: 394-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR