3-(4-Fluorophenyl)isoxazole-5-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A350594-1G A350594-250MG
77278-156EA 228.66 USD
77278-156 77748-687
3-(4-Fluorophenyl)isoxazole-5-carboxylic acid ≥98%
3-(4-Fluorophenyl)isoxazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆FNO₃
MDL Number: MFCD05150725
CAS Number: 618383-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR