3-Bromo-4-(trifluoromethoxy)aniline 98%

Supplier: Ambeed
Warning

A350292-100G A350292-10G A350292-25G
77278-150EA 83 USD
77278-150 77278-152 77278-154
3-Bromo-4-(trifluoromethoxy)aniline 98%
3-Bromo-4-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅BrF₃NO
MDL Number: MFCD00190127
CAS Number: 191602-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR