(Ir[Me(Me)ppy]2(dtbpy))PF6 97%

Supplier: Ambeed
Warning

A350160-1G A350160-250MG A350160-100MG
77251-526EA 715.85 USD
77251-526 77251-528 77572-024
(Ir[Me(Me)ppy]2(dtbpy))PF6 97%
(Ir[Me(Me)ppy]2(dtbpy))PF6

Precaución: for Research Use Only

Formula: C₄₄H₄₈F₆IrN₄P
MW: 970.05 g/mol
MDL Number: MFCD31707652
CAS Number: 1607469-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR