2-Benzylaniline 95%

Supplier: Ambeed
Warning

Synonyms: 2-Aminodiphenylmethane, 2-Benzylaniline

A349896-100G A349896-10G A349896-1G A349896-25G A349896-500G A349896-5G
77263-850EA 158.62 USD
77263-850 77263-852 77263-854 77263-856 77263-858 77263-860
2-Benzylaniline 95%
2-Benzylaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₃N
MW: 183.25 g/mol
MDL Number: MFCD00007750
CAS Number: 28059-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR