(S)-2-(1-Aminoethyl)-4-chlorophenol 97%

Supplier: Ambeed
Warning

A349632-250MG A349632-1G A349632-5G
77251-524EA 15.45 USD
77251-524 77746-687 77746-688
(S)-2-(1-Aminoethyl)-4-chlorophenol 97%
(S)-2-(1-Aminoethyl)-4-chlorophenol

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MW: 171.62 g/mol
MDL Number: MFCD06762060
CAS Number: 1228569-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR