2-Chloro-3-methoxybenzoic acid 97%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-m-anisic acid

A349513-100MG A349513-250MG A349513-5G A349513-1G
77263-844EA 7 USD
77263-844 77263-846 77263-848 77748-685
2-Chloro-3-methoxybenzoic acid 97%
2-Chloro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MDL Number: MFCD07784399
CAS Number: 33234-36-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR