2-(tert-Butylamino)ethyl methacrylate 98% stabilised

Supplier: Ambeed
Warning

Synonyms: Methacrylic acid 2-(tert-Butylamino)ethyl ester, 2-(tert-Butylamino)ethyl Methacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A349441-100G A349441-25G A349441-500G
77251-504EA 40 USD
77251-504 77251-506 77251-508
2-(tert-Butylamino)ethyl methacrylate 98% stabilised
2-(tert-Butylamino)ethyl methacrylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₂
MW: 185.27 g/mol
MDL Number: MFCD00014997
CAS Number: 3775-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR