AZ876 98+%

Supplier: Ambeed
Warning

A349245-10MG A349245-50MG A349245-5MG A349245-1MG
77263-836EA 50 USD
77263-836 77263-838 77263-840 77616-528
AZ876 98+%
AZ876

Precaución: for Research Use Only

Formula: C₂₄H₂₉N₃O₃S
MDL Number: MFCD30377191
CAS Number: 898800-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR