Ethyl 3-cyclobutyl-3-oxopropanoate 98%

Supplier: Ambeed
Warning

A349184-1G A349184-250MG A349184-5G
77278-138EA 18 USD
77278-138 77278-140 77748-684
Ethyl 3-cyclobutyl-3-oxopropanoate 98%
Ethyl 3-cyclobutyl-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₉H₁₄O₃
MDL Number: MFCD06410651
CAS Number: 24922-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR