5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Cyclohexyl N-Boc-L-glutamate, Boc-Glu(OcHex)-OH, N-(tert-Butoxycarbonyl)-L-glutamic Acid 5-Cyclohexyl Ester, N-Boc-L-glutamic Acid 5-Cyclohexyl Ester

A349127-100G A349127-10G A349127-1G A349127-25G
77278-130EA 92.7 USD
77278-130 77278-132 77278-134 77278-136
5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate 97%
5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate

Precaución: for Research Use Only

Formula: C₁₆H₂₇NO₆
MW: 329.39 g/mol
MDL Number: MFCD00065570
CAS Number: 73821-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR