2-Ethoxybenzoyl chloride 98%
Supplier: Ambeed
|
Danger
|
A349042-25G
A349042-5G
77263-832EA
18.54
USD
77263-832
77263-834
2-Ethoxybenzoyl chloride 98%
2-Ethoxybenzoyl chloride
Precaución: for Research Use Only
Formula:
C₉H₉ClO₂ MW: 184.62 g/mol |
MDL Number:
MFCD00016348 CAS Number: 42926-52-3 UN: 3265 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...