3-[3-(Trifluoromethyl)phenyl]-1-propanol 97%

Supplier: Ambeed
Warning

Synonyms: 3-(3-(Trifluoromethyl)phenyl)propan-1-ol

A348645-100G A348645-10G A348645-1G A348645-5G
77278-116EA 296.64 USD
77278-116 77278-118 77278-120 77278-122
3-[3-(Trifluoromethyl)phenyl]-1-propanol 97%
3-[3-(Trifluoromethyl)phenyl]-1-propanol

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₃O
MW: 204.19 g/mol
MDL Number: MFCD08706408
CAS Number: 78573-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR