3-(2,3,4-Trimethoxyphenyl)propionic acid 98+%

Supplier: Ambeed
Warning

Synonyms: 3-(2,3,4-Trimethoxyphenyl)propanoic acid

A348342-1G A348342-250MG A348342-5G
77278-110EA 50.47 USD
77278-110 77278-112 77278-114
3-(2,3,4-Trimethoxyphenyl)propionic acid 98+%
3-(2,3,4-Trimethoxyphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₅
MW: 240.26 g/mol
MDL Number: MFCD00002773
CAS Number: 33130-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR