Fmoc-Asp(OcHex)-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-L-aspartic acid 4-cyclohexyl ester

A347881-100G A347881-1G A347881-5G
77278-104EA 219.39 USD
77278-104 77278-106 77278-108
Fmoc-Asp(OcHex)-OH 98%
Fmoc-Asp(OcHex)-OH

Precaución: for Research Use Only

Formula: C₂₅H₂₇NO₆
MDL Number: MFCD00135420
CAS Number: 130304-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR