4-Hydroxy-N-phenylbenzenesulfonamide 98+%
Supplier: Ambeed
|
Warning
|
A347859-100G
A347859-25G
A347859-500G
77290-640EA
29
USD
77290-640
77290-642
77290-644
4-Hydroxy-N-phenylbenzenesulfonamide 98+%
4-Hydroxy-N-phenylbenzenesulfonamide
Precaución: for Research Use Only
Formula:
C₁₂H₁₁NO₃S MW: 249.28 g/mol |
MDL Number:
MFCD03840793 CAS Number: 161356-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...