trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: trans-3,4,5-Trifluoro-4'-(4-propylcyclohexyl)biphenyl, 3,4,5-Trifluoro-4'-(trans-4-propylcyclohexyl)biphenyl, 3,4,5-Trifluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

A347535-10G A347535-1G
77290-638EA 39 USD
77290-638 77748-681
trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl 98%
trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl

Precaución: for Research Use Only

Formula: C₂₁H₂₃F₃
MDL Number: MFCD09839001
CAS Number: 132123-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR