2-Cyanoethyl acrylate 96% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A347341-1G A347341-5G A347341-25G
77263-818EA 22 USD
77263-818 77263-820 77616-520
2-Cyanoethyl acrylate 96% stabilized
2-Cyanoethyl acrylate

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MDL Number: MFCD00013823
CAS Number: 106-71-8
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR