5-Chloro-2-fluoroaniline 98%

Supplier: Ambeed
Warning

A347011-100G A347011-10G A347011-25G A347011-500G A347011-5G
77290-628EA 55 USD
77290-628 77290-630 77290-632 77290-634 77290-636
5-Chloro-2-fluoroaniline 98%
5-Chloro-2-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅ClFN
MDL Number: MFCD00069416
CAS Number: 2106-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR