2-Bromo-3'-(trifluoromethyl)acetophenone 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-(3-(trifluoromethyl)phenyl)ethanone

A346907-100G A346907-10G A346907-1G A346907-5G
77278-086EA 1023.82 USD
77278-086 77278-088 77278-090 77572-010
2-Bromo-3'-(trifluoromethyl)acetophenone 98%
2-Bromo-3'-(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O
MW: 267.04 g/mol
MDL Number: MFCD03094283
CAS Number: 2003-10-3
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR