2-[1-(2-Thienyl)ethylidene]malononitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2-(1-(Thiophen-2-yl)ethylidene)malononitrile

A346567-1G A346567-5G
77251-488EA 40.17 USD
77251-488 77251-490
2-[1-(2-Thienyl)ethylidene]malononitrile 95%
2-[1-(2-Thienyl)ethylidene]malononitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂S
MW: 174.23 g/mol
MDL Number: MFCD00184599
CAS Number: 10432-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR