2'-Methylbiphenyl-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2'-Methyl-[1,1'-biphenyl]-3-carboxylic acid

A346225-1G A346225-250MG A346225-25G A346225-5G
77278-078EA 52.53 USD
77278-078 77278-080 77278-082 77278-084
2'-Methylbiphenyl-3-carboxylic acid 98%
2'-Methylbiphenyl-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
MDL Number: MFCD03424589
CAS Number: 168618-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR