1-Chloro-2,4-bis(trifluoromethyl)benzene 97%

Supplier: Ambeed
Danger

Synonyms: 1,3-Bis(trifluoromethyl)-4-chlorobenzene

A345838-10G A345838-1G A345838-25G A345838-5G
77251-474EA 71.07 USD
77251-474 77251-476 77251-478 77251-480
1-Chloro-2,4-bis(trifluoromethyl)benzene 97%
1-Chloro-2,4-bis(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₃ClF₆
MW: 248.56 g/mol
MDL Number: MFCD01631638
CAS Number: 327-76-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR