N-(5-Hydroxypyridin-2-yl)acetamide 98%

Supplier: Ambeed
Warning

A345796-100MG A345796-1G A345796-250MG A345796-5G
77251-466EA 53 USD
77251-466 77251-468 77251-470 77251-472
N-(5-Hydroxypyridin-2-yl)acetamide 98%
N-(5-Hydroxypyridin-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD09839685
CAS Number: 159183-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR