2-(Methylthio)pyrimidine-4,6(1H,5H)-dione 98%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxy-2-(methylthio)pyrimidin-4(3H)-one, 4,6-Dihydroxy-2-methylsulphanylpyrimidine, 4-Hydroxy-2-(methylthio)pyrimidin-6(1H)-one

A345437-100G A345437-10G A345437-25G A345437-500G A345437-5G
77290-582EA 53.56 USD
77290-582 77290-584 77290-586 77290-588 77290-590
2-(Methylthio)pyrimidine-4,6(1H,5H)-dione 98%
2-(Methylthio)pyrimidine-4,6(1H,5H)-dione

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂S
MW: 158.18 g/mol
MDL Number: MFCD00006087
CAS Number: 29639-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR