3,5-Di-tert-butyl-4-hydroxybenzenesulfonic acid 95%

Supplier: Ambeed
Warning

A345417-100MG A345417-250MG
77263-806EA 114.33 USD
77263-806 77263-808
3,5-Di-tert-butyl-4-hydroxybenzenesulfonic acid 95%
3,5-Di-tert-butyl-4-hydroxybenzenesulfonic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₂O₄S
MW: 286.39 g/mol
MDL Number: MFCD27965365
CAS Number: 25679-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR