3-Cyclohexyl-L-alanine hydrate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-cyclohexylpropanoic acid hydrate, (S)-(+)-a-Aminocyclohexanepropionic acid hydrate

A345215-100G A345215-10G A345215-25G A345215-5G
77278-070EA 138.02 USD
77278-070 77278-072 77278-074 77278-076
3-Cyclohexyl-L-alanine hydrate 97%
3-Cyclohexyl-L-alanine hydrate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂·H₂O
MW: 189.252 g/mol
MDL Number: MFCD03703498
CAS Number: 307310-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR