3-Ethoxy-4-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Apremilast Impurity 28

A344800-10G A344800-5G
77239-946EA 9.27 USD
77239-946 77239-948
3-Ethoxy-4-methoxybenzaldehyde 98%
3-Ethoxy-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
MDL Number: MFCD00010128
CAS Number: 1131-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR