(-)-Terpinen-4-ol 95%

Supplier: Ambeed
Warning

A344672-1G A344672-25G A344672-5G
77251-448EA 5 USD
77251-448 77251-450 77251-452
(-)-Terpinen-4-ol 95%
(-)-Terpinen-4-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₈O
MDL Number: MFCD00167108
CAS Number: 20126-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR