3-Fluoro-4-methylbenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-4-methylphenyl)methanol

A344615-100G A344615-1G A344615-25G A344615-5G
77278-062EA 635.51 USD
77278-062 77278-064 77278-066 77278-068
3-Fluoro-4-methylbenzyl alcohol 98%
3-Fluoro-4-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.16 g/mol
MDL Number: MFCD01631533
CAS Number: 192702-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR