6-Hydroxynicotinonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Formylphenyl)isonicotinonitrile, 6-Hydroxypyridine-3-carbonitrile, 5-Cyano-2-hydroxypyridine

A344441-100MG A344441-10G A344441-250MG A344441-25G A344441-5G
77278-052EA 9.27 USD
77278-052 77278-054 77278-056 77278-058 77278-060
6-Hydroxynicotinonitrile 98%
6-Hydroxynicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₄N₂O
MW: 120.109 g/mol
MDL Number: MFCD06656489
CAS Number: 95891-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR