3,4-Diaminothiophene dihydrochloride 95%

Supplier: Ambeed
Danger

A344291-100MG A344291-1G A344291-250MG
77278-046EA 9 USD
77278-046 77278-048 77278-050
3,4-Diaminothiophene dihydrochloride 95%
3,4-Diaminothiophene dihydrochloride

Precaución: for Research Use Only

Formula: C₄H₆N₂S
MDL Number: MFCD00671511
CAS Number: 90069-81-1
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR