1-Amino-1H-pyrrole-2-carboxamide 95%

Supplier: Ambeed
Warning

Synonyms: 1-Aminopyrrole-2-carboxamide

A344139-100MG A344139-250MG
77239-938EA 29.87 USD
77239-938 77239-940
1-Amino-1H-pyrrole-2-carboxamide 95%
1-Amino-1H-pyrrole-2-carboxamide

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.13 g/mol
MDL Number: MFCD11053506
CAS Number: 159326-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR