4-Methyl-m-anisic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxy-4-methylbenzoic acid

A343792-100G A343792-10G A343792-1G A343792-5G
77278-026EA 51.5 USD
77278-026 77278-028 77278-030 77278-032
4-Methyl-m-anisic acid 98%
4-Methyl-m-anisic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00002566
CAS Number: 7151-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR